Tisk…

2-(Cyclopentyloxy)acetic acid 95%

Dodavatel: Apollo Scientific

OR962786-1G OR962786-250MG OR962786-5G
APOSOR962786-1GEA 4300 CZK
APOSOR962786-1G APOSOR962786-250MG APOSOR962786-5G
2-(Cyclopentyloxy)acetic acid 95%
2-(Cyclopentyloxy)acetic acid
Vzorec: C₇H₁₂O₃
MW: 144.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09930082
CAS číslo: 95832-60-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR