Tisk…

4-(2-Methoxyethoxy)benzonitrile 98%

Dodavatel: Apollo Scientific

OR962771-1G OR962771-5G
APOSOR962771-1GEA 7280 CZK
APOSOR962771-1G APOSOR962771-5G
4-(2-Methoxyethoxy)benzonitrile 98%
4-(2-Methoxyethoxy)benzonitrile
Vzorec: C₁₀H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD09907643
CAS číslo: 80407-66-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR