Isopropyl-(2-nitro-phenyl)-amine 95%
Dodavatel: Apollo Scientific
OR962765-1G
OR962765-5G
APOSOR962765-1GEA
7050
CZK
APOSOR962765-1G
APOSOR962765-5G
Isopropyl-(2-nitro-phenyl)-amine 95%
Isopropyl-(2-nitro-phenyl)-amine
Vzorec:
C₉H₁₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09862609 CAS číslo: 25186-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...