Tisk…

2-(Isopropylthio)aniline 95%

Dodavatel: Apollo Scientific

OR962705-10G OR962705-1G OR962705-5G
APOSOR962705-10GEA 4600 CZK
APOSOR962705-10G APOSOR962705-1G APOSOR962705-5G
2-(Isopropylthio)aniline 95%
2-(Isopropylthio)aniline
Vzorec: C₉H₁₃NS
MW: 167,271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08691961
CAS číslo: 6397-33-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR