2-Methylquinoline-4-carboxaldehyde 98%
Dodavatel: Apollo Scientific
OR962689-100MG
OR962689-1G
OR962689-250MG
APOSOR962689-100MGEA
4140
CZK
APOSOR962689-100MG
APOSOR962689-1G
APOSOR962689-250MG
2-Methylquinoline-4-carboxaldehyde 98%
2-Methylquinoline-4-carboxaldehyde
Vzorec:
C₁₁H₉NO MW: 171,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08052651 CAS číslo: 6760-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...