3-(2-Hydroxy-ethoxy)-benzoic acid 95%
Dodavatel: Apollo Scientific
OR962645-1G
OR962645-250MG
APOSOR962645-1GEA
10350
CZK
APOSOR962645-1G
APOSOR962645-250MG
3-(2-Hydroxy-ethoxy)-benzoic acid 95%
3-(2-Hydroxy-ethoxy)-benzoic acid
Vzorec:
C₉H₁₀O₄ MW: 182,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203193 CAS číslo: 25781-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...