(4-Pyrrolidin-1-ylphenyl)methanol 95%
Dodavatel: Apollo Scientific
OR962644-1G
OR962644-25G
OR962644-5G
APOSOR962644-1GEA
4140
CZK
APOSOR962644-1G
APOSOR962644-25G
APOSOR962644-5G
(4-Pyrrolidin-1-ylphenyl)methanol 95%
(4-Pyrrolidin-1-ylphenyl)methanol
Vzorec:
C₁₁H₁₅NO MW: 177,243 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203114 CAS číslo: 676245-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...