(5-Methylthiophen-2-yl)methanol 97%
Dodavatel: Apollo Scientific
OR962642-1G
OR962642-5G
APOSOR962642-1GEA
2280
CZK
APOSOR962642-1G
APOSOR962642-5G
(5-Methylthiophen-2-yl)methanol 97%
(5-Methylthiophen-2-yl)methanol
Vzorec:
C₆H₈OS MW: 128,192 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06202627 CAS číslo: 63826-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...