(4-Piperidin-1-yl-phenyl)methanol 98%
Dodavatel: Apollo Scientific
OR962640-1G
OR962640-25G
OR962640-5G
APOSOR962640-1GEA
3740
CZK
APOSOR962640-1G
APOSOR962640-25G
APOSOR962640-5G
(4-Piperidin-1-yl-phenyl)methanol 98%
(4-Piperidin-1-yl-phenyl)methanol
Vzorec:
C₁₂H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD05864822 CAS číslo: 677764-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...