2-(Imidazol-1-yl)benzaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR962635-1G
OR962635-25G
OR962635-5G
APOSOR962635-1GEA
3110
CZK
APOSOR962635-1G
APOSOR962635-25G
APOSOR962635-5G
2-(Imidazol-1-yl)benzaldehyde 97%
2-(Imidazol-1-yl)benzaldehyde
Vzorec:
C₁₀H₈N₂O MW: 172,183 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05864671 CAS číslo: 151055-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...