5-Methyl-1H-pyrrole-2-carboxylic acid 98%
Dodavatel: Apollo Scientific
OR962602-1G
OR962602-5G
APOSOR962602-1GEA
4770
CZK
APOSOR962602-1G
APOSOR962602-5G
5-Methyl-1H-pyrrole-2-carboxylic acid 98%
5-Methyl-1H-pyrrole-2-carboxylic acid
Vzorec:
C₆H₇NO₂ MW: 125,125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03011381 CAS číslo: 3757-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...