Tisk…

N-Ethyl-4-nitrobenzamide 98%

Dodavatel: Apollo Scientific

OR962553-1G OR962553-5G
APOSOR962553-1GEA 5200 CZK
APOSOR962553-1G APOSOR962553-5G
N-Ethyl-4-nitrobenzamide 98%
N-Ethyl-4-nitrobenzamide
Vzorec: C₉H₁₀N₂O₃
MW: 194,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01211872
CAS číslo: 50445-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR