Tisk…
N,N-Diethyl-o-nitroaniline 98%
Dodavatel: Apollo Scientific
OR962549-1G OR962549-5G
APOSOR962549-1GEA 6220 CZK
APOSOR962549-1G APOSOR962549-5G
N,N-Diethyl-o-nitroaniline 98%
N,N-Diethyl-o-nitroaniline
Vzorec: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01120260
CAS číslo: 2216-17-3