Tisk…

4-(2-Hydroxyethoxy)benzonitrile 96%

Dodavatel: Apollo Scientific

OR962538-1G OR962538-5G
APOSOR962538-1GEA 5720 CZK
APOSOR962538-1G APOSOR962538-5G
4-(2-Hydroxyethoxy)benzonitrile 96%
4-(2-Hydroxyethoxy)benzonitrile
Vzorec: C₉H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00832129
CAS číslo: 57928-96-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR