Tisk…

N-Butyl-4-nitroaniline 98%

Dodavatel: Apollo Scientific

OR962453-5G
APOSOR962453-5GEA 1780 CZK
APOSOR962453-5G
N-Butyl-4-nitroaniline 98%
N-Butyl-4-nitroaniline
Vzorec: C₁₀H₁₄N₂O₂
MW: 194,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024635
CAS číslo: 58259-34-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR