Tisk…

2-(4-Chlorophenyl)pyridine 95%

Dodavatel: Apollo Scientific

OR962436-1G
APOSOR962436-1GEA 2240 CZK
APOSOR962436-1G
2-(4-Chlorophenyl)pyridine 95%
2-(4-Chlorophenyl)pyridine
Vzorec: C₁₁H₈ClN
MW: 189,641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201380
CAS číslo: 5969-83-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR