Tisk…

2-(3-Chlorophenyl)pyrrolidine 97%

Dodavatel: Apollo Scientific

OR962418-1G OR962418-5G
APOSOR962418-1GEA 4770 CZK
APOSOR962418-1G APOSOR962418-5G
2-(3-Chlorophenyl)pyrrolidine 97%
2-(3-Chlorophenyl)pyrrolidine
Vzorec: C₁₀H₁₂ClN
Storage Temperature: Ambient
MDL Number: MFCD02663429
CAS číslo: 298690-74-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR