Tisk…

2-(3-Chlorophenyl)acetamide 95%

Dodavatel: Apollo Scientific

OR962416-1G OR962416-250MG
APOSOR962416-1GEA 857 CZK
APOSOR962416-1G APOSOR962416-250MG
2-(3-Chlorophenyl)acetamide 95%
2-(3-Chlorophenyl)acetamide
Vzorec: C₈H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD09991645
CAS číslo: 58357-84-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR