Tisk…

Methyl 2-Hydroxycyclopentylcarboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-hydroxycyclopentanecarboxylate

OR962411-1G OR962411-25G OR962411-5G
APOSOR962411-1GEA 1540 CZK
APOSOR962411-1G APOSOR962411-25G APOSOR962411-5G
Methyl 2-Hydroxycyclopentylcarboxylate 97%
Methyl 2-Hydroxycyclopentylcarboxylate
Vzorec: C₇H₁₂O₃
MW: 144,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972392
CAS číslo: 90085-05-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR