6-Methyl-chroman-4-ylamine hydrochloride 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR962407-250MG
OR962407-1G
APOSOR962407-250MGEA
10350
CZK
APOSOR962407-250MG
APOSOR962407-1G
6-Methyl-chroman-4-ylamine hydrochloride 95+%
6-Methyl-chroman-4-ylamine hydrochloride
Vzorec:
C₁₀H₁₃NO·ClH Storage Temperature: Refrigerator |
MDL Number:
MFCD09026798 CAS číslo: 191608-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...