3-Propoxy-azetidine hydrochloride 98%
Dodavatel: Apollo Scientific
Synonyms:
3-Propoxyazetidine hydrochloride
OR962398-1G
OR962398-5G
APOSOR962398-1GEA
20830
CZK
APOSOR962398-1G
APOSOR962398-5G
3-Propoxy-azetidine hydrochloride 98%
3-Propoxy-azetidine hydrochloride
Vzorec:
C₆H₁₃NO·ClH MW: 151,634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06804557 CAS číslo: 897019-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...