Tisk…

4-(2-Methoxy-ethoxy)-phenol 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(2-Methoxyethoxy)phenol

OR962223-1G OR962223-250MG
APOSOR962223-1GEA 2480 CZK
APOSOR962223-1G APOSOR962223-250MG
4-(2-Methoxy-ethoxy)-phenol 95%
4-(2-Methoxy-ethoxy)-phenol
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114941
CAS číslo: 51980-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR