(S)-2-Amino-5-phenylpent-4-enoic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
H-Styryl-Ala-OH
OR962051-1G
APOSOR962051-1GEA
6290
CZK
APOSOR962051-1G
(S)-2-Amino-5-phenylpent-4-enoic acid 97%
(S)-2-Amino-5-phenylpent-4-enoic acid
Vzorec:
C₁₁H₁₃NO₂ MW: 191,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01860887 CAS číslo: 267650-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...