Tisk…

3-(5-Bromothien-2-yl)-L-alanine 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-2-Amino-3-(5-bromothiophen-2-yl)propanoic acid, H-2-(5-Br-Thienyl)-Ala-OH

OR962048-1G
APOSOR962048-1GEA 3770 CZK
APOSOR962048-1G
3-(5-Bromothien-2-yl)-L-alanine 97%
3-(5-Bromothien-2-yl)-L-alanine
Vzorec: C₇H₈BrNO₂S
MW: 250,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117834
CAS číslo: 154593-58-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR