Tisk…

Quinoline-8-thiocarboxamide 95%

Dodavatel: Apollo Scientific

OR961932-1G OR961932-250MG
APOSOR961932-1GEA 22950 CZK
APOSOR961932-1G APOSOR961932-250MG
Quinoline-8-thiocarboxamide 95%
Quinoline-8-thiocarboxamide
Vzorec: C₁₀H₈N₂S
MW: 188,249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738915
CAS číslo: 62216-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR