Tisk…

2-((Pyrrolidin-1-yl)methyl) morpholine 95%

Dodavatel: Apollo Scientific

OR961779-100MG OR961779-1G OR961779-250MG
APOSOR961779-100MGEA 4570 CZK
APOSOR961779-100MG APOSOR961779-1G APOSOR961779-250MG
2-((Pyrrolidin-1-yl)methyl) morpholine 95%
2-((Pyrrolidin-1-yl)methyl) morpholine
Vzorec: C₉H₁₈N₂O
MW: 170,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658512
CAS číslo: 128208-00-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR