Tisk…

Sodium pivalate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Sodium 2,2-dimethylpropanoate

OR961732-25G
APOSOR961732-25GEA 2360 CZK
APOSOR961732-25G
Sodium pivalate 95%
Sodium pivalate
Vzorec: C₅H₉O₂·Na
MW: 124,114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064202
CAS číslo: 1184-88-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR