4-(4-Hydroxyphenyl)piperazine, N1-BOC protected 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl 4-(4-hydroxyphenyl)piperazine-1-carboxylate
OR961614-25G
OR961614-5G
APOSOR961614-25GEA
11410
CZK
APOSOR961614-25G
APOSOR961614-5G
4-(4-Hydroxyphenyl)piperazine, N1-BOC protected 97%
4-(4-Hydroxyphenyl)piperazine, N1-BOC protected
Vzorec:
C₁₅H₂₂N₂O₃ MW: 278,347 g/mol Bod tání: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115060 CAS číslo: 158985-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...