Tisk…

2-Cycloheptyl-ethylamine 95%

Dodavatel: Apollo Scientific

OR961602-100MG OR961602-1G OR961602-250MG
APOSOR961602-100MGEA 5200 CZK
APOSOR961602-100MG APOSOR961602-1G APOSOR961602-250MG
2-Cycloheptyl-ethylamine 95%
2-Cycloheptyl-ethylamine
Vzorec: C₉H₁₉N
Storage Temperature: Ambient
MDL Number: MFCD04114510
CAS číslo: 4448-84-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR