Tisk…
1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
Dodavatel: Apollo Scientific
Synonyms: 1-Amino-3,3-dimethylbutan-2-one hydrochloride
OR961587-1G OR961587-5G
APOSOR961587-1GEA 827 CZK
APOSOR961587-1G APOSOR961587-5G
1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
1-Amino-3,3-dimethyl-butan-2-one hydrochloride
Vzorec: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798286
CAS číslo: 33119-72-1