Tisk…

Fmoc-(R)-3-Cyclobutylalanine 97%

Dodavatel: Apollo Scientific

OR961533-1G OR961533-250MG
APOSOR961533-1GEA 1160 CZK
APOSOR961533-1G APOSOR961533-250MG
Fmoc-(R)-3-Cyclobutylalanine 97%
Fmoc-(R)-3-Cyclobutylalanine
Vzorec: C₂₂H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD03840401
CAS číslo: 478183-63-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR