4-(Methoxymethoxy)benzoic acid 95%
Dodavatel: Apollo Scientific
OR961500-1G
OR961500-5G
APOSOR961500-1GEA
705
CZK
APOSOR961500-1G
APOSOR961500-5G
4-(Methoxymethoxy)benzoic acid 95%
4-(Methoxymethoxy)benzoic acid
Vzorec:
C₉H₁₀O₄ MW: 182,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21101751 CAS číslo: 25458-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...