Tisk…

(3,5-Dichloro-phenyl)-ethyl-amine 95%

Dodavatel: Apollo Scientific

OR961415-1G OR961415-250MG
APOSOR961415-1GEA 17990 CZK
APOSOR961415-1G APOSOR961415-250MG
(3,5-Dichloro-phenyl)-ethyl-amine 95%
(3,5-Dichloro-phenyl)-ethyl-amine
Vzorec: C₈H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD03210688
CAS číslo: 42266-02-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR