(3,4-Dichloro-phenyl)-ethyl-amine 95%
Dodavatel: Apollo Scientific
OR961414-1G
OR961414-250MG
APOSOR961414-1GEA
12400
CZK
APOSOR961414-1G
APOSOR961414-250MG
(3,4-Dichloro-phenyl)-ethyl-amine 95%
(3,4-Dichloro-phenyl)-ethyl-amine
Vzorec:
C₈H₉Cl₂N MW: 190.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03210687 CAS číslo: 17847-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...