(S)-3-Amino-4-(4-chlorophenyl)butanoic acid hydrochloride
Dodavatel: Apollo Scientific
OR961272-1G
OR961272-250MG
OR961272-5G
APOSOR961272-1GEA
8070
CZK
APOSOR961272-1G
APOSOR961272-250MG
APOSOR961272-5G
(S)-3-Amino-4-(4-chlorophenyl)butanoic acid hydrochloride
(S)-3-Amino-4-(4-chlorophenyl)butanoic acid hydrochloride
Vzorec:
C₁₀H₁₂ClNO₂·ClH Bod tání: 210…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861001 CAS číslo: 331763-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...