Tisk…

(R)-2-Amino-5-phenylpent-4-enoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: H-D-Styryl-Ala-OH

OR961251-1G
APOSOR961251-1GEA 3310 CZK
APOSOR961251-1G
(R)-2-Amino-5-phenylpent-4-enoic acid 97%
(R)-2-Amino-5-phenylpent-4-enoic acid
Vzorec: C₁₁H₁₃NO₂
MW: 191,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860880
CAS číslo: 264903-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR