Tisk…

Boc-4-Acetyl-L-phenylalanine 97%

Dodavatel: Apollo Scientific

OR961228-1G OR961228-500MG
APOSOR961228-1GEA 11180 CZK
APOSOR961228-1G APOSOR961228-500MG
Boc-4-Acetyl-L-phenylalanine 97%
Boc-4-Acetyl-L-phenylalanine
Vzorec: C₁₆H₂₁NO₅
MW: 307,342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632035
CAS číslo: 204856-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR