Tisk…

4,4-Dimethyl-2-oxo-pentanoic acid 95%

Dodavatel: Apollo Scientific

OR961208-1G OR961208-5G
APOSOR961208-1GEA 5390 CZK
APOSOR961208-1G APOSOR961208-5G
4,4-Dimethyl-2-oxo-pentanoic acid 95%
4,4-Dimethyl-2-oxo-pentanoic acid
Vzorec: C₇H₁₂O₃
MW: 144,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01325393
CAS číslo: 34906-87-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR