Tisk…

1-(1-Methylpyrazol-4-yl)ethanol 95%

Dodavatel: Apollo Scientific

OR960930-100MG OR960930-1G OR960930-250MG
APOSOR960930-100MGEA 4570 CZK
APOSOR960930-100MG APOSOR960930-1G APOSOR960930-250MG
1-(1-Methylpyrazol-4-yl)ethanol 95%
1-(1-Methylpyrazol-4-yl)ethanol
Vzorec: C₆H₁₀N₂O
MW: 126.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08700894
CAS číslo: 40534-33-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR