Tisk…

Chloromethyl benzoate 98%

Dodavatel: Apollo Scientific

OR960834-10G OR960834-25G
APOSOR960834-10GEA 3940 CZK
APOSOR960834-10G APOSOR960834-25G
Chloromethyl benzoate 98%
Chloromethyl benzoate
Vzorec: C₈H₇ClO₂
MW: 170,593 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040116
CAS číslo: 5335-05-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR