2-[(Pyrrolidin-1-yl)carbonyl]pyridine 97%
Dodavatel: Apollo Scientific
OR960341-1G
OR960341-5G
APOSOR960341-1GEA
7670
CZK
APOSOR960341-1G
APOSOR960341-5G
2-[(Pyrrolidin-1-yl)carbonyl]pyridine 97%
2-[(Pyrrolidin-1-yl)carbonyl]pyridine
Vzorec:
C₁₀H₁₂N₂O MW: 176,215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10652053 CAS číslo: 83728-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...