tert-Butyl N-(1-cyanoethyl)carbamate 98%
Dodavatel: Apollo Scientific
OR960329-1G
OR960329-250MG
APOSOR960329-1GEA
12400
CZK
APOSOR960329-1G
APOSOR960329-250MG
tert-Butyl N-(1-cyanoethyl)carbamate 98%
tert-Butyl N-(1-cyanoethyl)carbamate
Vzorec:
C₈H₁₄N₂O₂ MW: 170.209 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12792276 CAS číslo: 141041-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...