Tisk…

tert-Butyl N,N-dimethylcarbamate 98%

Dodavatel: Apollo Scientific

OR960324-1G OR960324-5G
APOSOR960324-1GEA 5200 CZK
APOSOR960324-1G APOSOR960324-5G
tert-Butyl N,N-dimethylcarbamate 98%
tert-Butyl N,N-dimethylcarbamate
Vzorec: C₇H₁₅NO₂
MW: 145,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318374
CAS číslo: 7541-17-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR