Tisk…

3-Chloro-4-formylbenzoic acid 95%

Dodavatel: Apollo Scientific

OR960323-1G OR960323-250MG
APOSOR960323-1GEA 9200 CZK
APOSOR960323-1G APOSOR960323-250MG
3-Chloro-4-formylbenzoic acid 95%
3-Chloro-4-formylbenzoic acid
Vzorec: C₈H₅ClO₃
Storage Temperature: Ambient
MDL Number: MFCD20649616
CAS číslo: 58588-59-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR