Tisk…

3-(3-Chlorophenyl)-1,2,4-triazole 98%

Dodavatel: Apollo Scientific

OR960306-1G OR960306-5G
APOSOR960306-1GEA 4770 CZK
APOSOR960306-1G APOSOR960306-5G
3-(3-Chlorophenyl)-1,2,4-triazole 98%
3-(3-Chlorophenyl)-1,2,4-triazole
Vzorec: C₈H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD24394207
CAS číslo: 52853-74-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR