Tisk…

3-(2-Chlorophenyl)-4H-1,2,4-triazole 98%

Dodavatel: Apollo Scientific

OR960305-1G OR960305-5G
APOSOR960305-1GEA 6220 CZK
APOSOR960305-1G APOSOR960305-5G
3-(2-Chlorophenyl)-4H-1,2,4-triazole 98%
3-(2-Chlorophenyl)-4H-1,2,4-triazole
Vzorec: C₈H₆ClN₃
MW: 179.606 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07352278
CAS číslo: 56015-92-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR