5-Bromothiophen-2-amine, N-BOC protected 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
tert-Butyl (5-bromothiophen-2-yl)carbamate
OR960289-1G
OR960289-5G
APOSOR960289-1GEA
2980
CZK
APOSOR960289-1G
APOSOR960289-5G
5-Bromothiophen-2-amine, N-BOC protected 96%
5-Bromothiophen-2-amine, N-BOC protected
Vzorec:
C₉H₁₂BrNO₂S MW: 278,166 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06659527 CAS číslo: 943321-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...