Tisk…

4-Cyano-2-methylbenzoic acid 96%

Dodavatel: Apollo Scientific
Varování

OR960287-10G OR960287-1G OR960287-5G
APOSOR960287-10GEA 5360 CZK
APOSOR960287-10G APOSOR960287-1G APOSOR960287-5G
4-Cyano-2-methylbenzoic acid 96%
4-Cyano-2-methylbenzoic acid
Vzorec: C₉H₇NO₂
MW: 161,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14582960
CAS číslo: 1975-53-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR