Tisk…
Pent-4-en-1-amine hydrochloride 96%
Dodavatel: Apollo Scientific
OR960248-1G OR960248-5G
APOSOR960248-1GEA 6250 CZK
APOSOR960248-1G APOSOR960248-5G
Pent-4-en-1-amine hydrochloride 96%
Pent-4-en-1-amine hydrochloride
Vzorec: C₅H₁₁N·ClH
Storage Temperature: Ambient
MDL Number: MFCD08437626
CAS číslo: 27546-60-7