Methyl 1-methylcyclohexane-1-carboxylate 98%
Dodavatel: Apollo Scientific
OR960236-1G
OR960236-5G
APOSOR960236-1GEA
5200
CZK
APOSOR960236-1G
APOSOR960236-5G
Methyl 1-methylcyclohexane-1-carboxylate 98%
Methyl 1-methylcyclohexane-1-carboxylate
Vzorec:
C₉H₁₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12025265 CAS číslo: 825-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...